Learn More
Labconco Vacuum Sensor for FreeZone
Contact Labconco's Service Departement for more information.
Brand:  Labconco 7445400
Additional Details : Weight : 0.20000kg
Product Code. 15175045
  • € 188.00 / Each
Estimated Shipment: 02-10-2024
to see stock.
Add to basket
Description
Description
Replacement Vacuum Sensor for all FreeZone Freeze Dryers made after 2004 (Also need 3 digit calibration code from side of sensor). Contact Labconco's Service Departement for more information.
Specifications
Specifications
1.0 lb.
0.5 kg