Learn More
Kirsch Philipp™ Labex™ Laboratory Refrigerator
Features explosion proof interior.
Brand: Kirsch Philipp™ 10315
Additional Details : Weight : 30.00000kg
Description
- Explosion proof interior
- Automatic defrosting
- Meltwater tray for manual emptying
- Easy-to-read LED temperature display
- Easy accessible control panel with key switch
- Micro-processor controlled temperature, thawing regulation to maximize temperature stability
- Minimum/maximum temperature memory
- Safety device to prevent freezing
- Optical and acoustical alarm
- RS-485 interface for computer based temperature documentation
- Potential free contact for alarm transmission
Specifications
Laboratory Freezer | |
2°C to 20°C | |
16 to 40 kg | |
Four (height adjustable) shelves, two drawers | |
50/60 Hz | |
0.9kWh/24 hr. |
330 L | |
6 Adjustable | |
Storage of laboratory chemicals | |
45 x 53x 128 cm | |
120 w | |
88 kg( gross) |