Learn More
Carl Roth™ Microwave
Brand:  Carl Roth™ HX94.1
Additional Details : Weight : 0.01000kg
Product Code. 10372800
  • POA / Each
Estimated Shipment: 20-08-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Stainless Steel
Up to 90 min.
375 mm
395 mm
230 V, 50 Hz
Large-capacity microwave oven
395 x 375 x 272 mm
272 mm
approx. 21 kg
1050 w