Learn More
RETSCH MM 400 Mixer Mills: Grinding Balls
For use with MM 400 Mixer Mills
Brand:  RETSCH 05.368.0093
Additional Details : Weight : 0.02500kg
 View more versions of this product
Product Code. 10324771
  • € 133.00 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
Various sizes and materials.
Specifications
Specifications
Grinding Ball
2 cm
MM 400 mixer mill
0.78 in.
Ideal for increasing the degree of mixing, grinding
Zirconium Oxide