Learn More
Invitrogen™ NA-XTD™ Influenza Neuraminidase Assay Reagent Set, no plates included
Next-generation chemiluminescence-based assay for long-lived emission kinetics for quantitation of influenza neuraminidase inhibitor resistance. Thermo Scientific NA-XTD Influenza Neuraminidase Assay Kit can be used for virus quantitation in media samples
Brand: Invitrogen™ 4457534
1748.00 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.87000kg
Includes: Influenza Neuraminidase Assay Kit includes 1000X NA-XTD Substrate, NA Sample Prep Buffer, NA-XTD Assay Buffer, NA-XTD Accelerator and 10 detection microplates Influenza Neuraminidase Assay Reagent Set includes 1000X NA-XTD Substrate, NA Sample Prep Buffer
Description
- Superior performance - improved, long-lived light emission kinetics and read-time flexibility
- High sensitivity - high assay signal-to-noise, detection of low-virus concentrations, and wide assay dynamic range
- Simple instrumentation requirement - no reagent injectors or luminometer needed
- Multiple applications - neuraminidase inhibitor IC50 assays and cell-based virus quantitation
- Detection Method: Chemiluminescent
- For Use With Equipment: Luminometer (Microplate)
- Form: Liquid
- Format: 96-well plate
- High Throughput Compatibility: High Throughput-Compatible
- Readout: End Point
- Substrate: NA-XTD
- Substrate Properties: Chemical Substrate
- Substrate Type: Neuraminidase Substrate
- Target Enzyme: Neuraminidase, Sialidase
Specifications
This kit contains 1000X NA-XTD™ Substrate, NA Sample Prep Buffer, NA-XTD™ Assay Buffer, and NA-XTD™ Accelerator. Store all components at 2°C to 8°C. | |
Influenza Neuraminidase Assay | |
Neuraminidase Assay | |
96-well plate | |
NA-XTD™, NovaBright™ | |
NA-XTD™ | |
Neuraminidase Substrate | |
High-throughput Compatible |
Enhanced Chemiluminescence | |
Luminometer (Microplate) | |
Liquid | |
Less hazardous, Sustainable packaging | |
Wet Ice | |
Chemical Substrate | |
Sialidase, Neuraminidase | |
Enzyme Assay |
For Research Use Only. Not for use in diagnostic procedures.