Learn More
NUMB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132452
Additional Details : Weight : 0.01000kg
Description
PA1-32452 detects Numb from mouse, human samples. PA1-32452 has been successfully used in Western blot applications. The PA1-32452 immunogen is: Synthetic peptide corresponding to a C-terminal region of the mouse numb protein sequence (between residues 600-653).
The protein encoded by this gene plays a role in the determination of cell fates during development. The encoded protein, whose degradation is induced in a proteasome-dependent manner by MDM2, is a membrane-bound protein that has been shown to associate with EPS15, LNX1, and NOTCH1. Four transcript variants encoding different isoforms have been found for this gene.Specifications
NUMB | |
Polyclonal | |
Unconjugated | |
P49757, Q9QZS3, Q2LC84 | |
NUMB | |
Synthetic peptide corresponding to a C-terminal region of the murine numb protein sequence (between residues 600-653). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
3.5 mg/mL | |
NUMB | |
S171 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18222, 29419, 8650 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |