Learn More
DWK Life Sciences O-Ring, suitable only for desiccator Base with ring-grooved flange, from silicone (VMQ)
The O-ring is a seal suitable for the desiccator with fluted bevel and enables a tight seal.
Brand: DWK Life Sciences 292156108
31.49 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.02000kg
Description
The O-ring is a seal suitable for the desiccator with fluted bevel, which seals the desiccator tightly with a precisely fitting shape. The use of the seal eliminates the need for grinding grease.
- With the help of the O-ring, a tight seal of the desiccator with annular groove flange can be achieved
- Suitable only for desiccator base with ring-grooved flange
Specifications
Tight closure of desiccators; Ordering advice: the O-ring is dimensioned according to the nominal doameter (DN) | |
Silicone | |
For Models From 1996 |
1 Ea. | |
236 mm |