Learn More
Heidolph Instruments Reax 2 Overhead Shaker
Compact for small volumes
Brand:  Heidolph Instruments 541-21001-00
Additional Details : Weight : 4.80000kg
Product Code. 16382106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Designed for simultaneous mixing of up to 20 vessels with diameters of 10 to 18 mm
    • Corresponds to the size of classic test tubes or 15 mL Falcon™ tubes
  • Alternatively, you can insert your complete rack into the universal adapter (width: 230 mm, height min./max.: 45 mm/160 mm) of the overhead shaker
  • Equipped with a solid metal frame, its design stands for extreme load capacity and reliability
  • Guaranteed safety during unattended continuous operation
Specifications
Specifications
Adjustable
230 V
Universal Adaptor allows for the use of Various Vessels of 50 to 160mm Height
20 to 100 rpm
Overhead Shaker