Learn More
3M™ Paper Sheet 618, P240
3M™ Paper Sheet 618 is a silicon carbide mineral abrasive on a flexible backing with a Free-Cut anti-loading coating.
Brand: 3M™ 7000034040
Additional Details : Weight : 1.00000kg
Beschreibung
- Provides a sharp, aggressive cut with a fine finish due to the silicon carbide abrasive mineral
- Features a Free-Cut anti-loading coating
- Contains a flexible backing
- Hand Blocks and Randon Orbital Sanders
- Size: 230 x 280 mm
Spezifikation
Paper |