Learn More
Bohlender™ PMMA SICCO Star Horizontal Desiccator
Ideal for storing humidity-sensitive products using silica gel. Bohlender™ PMMA SICCO Star Horizontal Desiccator is designed with a tightly fitting door, a reinforced aluminum frame, and clear surrounding panels.
Brand: Bohlender™ V1899-07
Additional Details : Weight : 8.24000kg
Description
- Uses silica gel.
- Tightly fitting door protects the contents from atmospheric contamination.
- Controlled environment is ideal for storing reference materials, library samples, metallographic specimens, tobacco and DNA-samples.
- Reinforced aluminum frame with acrylic panels.
- Door with magnetic catch and circular rubber seal.
- Non-slip rubber feet.
- Easy-to-read electronic digital hygrometer with temperature and humidity display.
- Variable height shelves made of acrylic glass.
- Telescopic shelves.
- Desiccant tray.
- Silica gel included.
- Raster and rails made metal-free PA/fiberglass.
- Rails with level numbering.
- Can be equipped with up to 13 shelves.
- Raster and rails made metal-free PA/fiberglass.
- Rails with level numbering.
Specifications
Clear | |
Horizontal Desiccator Cabinet | |
Telescopic | |
320 x 480 x 260 mm | |
480 mm | |
260 mm | |
320 mm |
PMMA | |
1 Each | |
42 L | |
525 mm | |
340 mm | |
375 mm | |
7.0 kg |