missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polystyrene Non-sterile Pipets

Brand:  VITLAB™ 160519

Additional Details : Weight : 0.09300kg

 View more versions of this product

Product Code. 15261549

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Polystyrene
Yes
10 Pack
5 mL
Pipet
Non-sterile
0.10 mL
Transparent
320 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers