Learn More
Edmund Bühler GmbH™ Power Control LR 1
Provides stepless control of the current consumption between 0 to 100%. Edmund Bühler GmbH™ Power Control LR 1 is for use with IRB 1 Infrared Heater.
Brand:  Edmund Bühler GmbH™ 6070 000
Additional Details : Weight : 100.00000kg
Product Code. 10160761
  • € 338.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • For use with IRB 1 Infrared Heater
  • Provides stepless control of current consumption from 0 to 100%.
Specifications
Specifications
IRB 1 Infrared Heater
Power Control LR 1