missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Vacuum Filter Flask with Screw Thread Connector And Ground Socket
Complies with ISO 6556
Brand: Quickfit FB100/2SC
437.00 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.30000kg
Description
- Complies with ISO 6556
- Büchner flasks with precision-ground Quickfit™ socket
- Side-arm features a PTFE screwthread connector for the safe attachment of 9mm bore vacuum tubing; Unscrew connector from flask and fit tubing, rescrew connector and tubing to flask
- Heavy walls and bases for vacuum filtration work
Specifications
ISO 6556 | |
Borosilicate Glass | |
PTFE screw thread connector |
100 mL | |
107 mm | |
64 mm |