missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 733 for Human FLRG
For use with catalog number DFLRG0
Brand: R&D Systems™ QC91
Additional Details : Weight : 0.00970kg
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
10272 | |
1 Each |
Follistatin-related Gene Protein/FLRG | |
Control | |
Human |