Learn More
Thermo Scientific™ Rapid Trehalose Assimilation Broth
Product Code. 10691345
Change view
Click to view available options
Quantity:
20 x 2 mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10691345 20 x 2 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10691345 Supplier Thermo Scientific™ Supplier No. R064856
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Rapidly identify Candida (Torulopsis) glabrata using Rapid Trehalose Assimilation Broth.
Rapidly identify Candida (Torulopsis) glabrata using Thermo Scientific™ Remel™ Rapid Trehalose Assimilation Broth. In 1985, Stockman and Roberts described the use of rapid assimilation trehalose broth to screen germ-tube-negative yeast isolates for the presumptive identification of C. glabrata1. The test was performed in a microtiter format and interpreted after 1 hour incubation at 35° C. The format and formulation of this screening method have been modified to accommodate the use of a tubed medium.
Candida glabrata represents approximately 20% of all yeast recovered from urine cultures2. Recently, C. glabrata has emerged as an opportunistic pathogen with cases of endocarditis, meningitis and disseminated infections.
  • Ready to use – Convenience of a prepared media
  • Rapid results – Results are obtained within 1-3 hours
  • Easy to identify – Bromcresol green dye turns to yellow to yellow-green color development shows a positive result
  • The incubation temperature is increased to 42°C to improve specificity and to facilitate rapid results
This medium contains higher amounts of yeast nitrogen and carbohydrate (trehalose) compared to the Wickerham broth formulation. Bromocresol green is incorporated as an indicator because its lower pH range is active in a more acidic environment.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Rapid Trehalose Assimilation Broth
Format 12mm x 75mm Tube
Product Type Microbiology Media
Quantity 20 x 2 mL