Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
Vapor duct with Combi-Clip for use with connecting the evaporating flask to the Rotavapor.
Brand:  BUCHI 11062187
 View more versions of this product
Product Code. 17603135
  • € 159.00 / Each
Estimated Shipment: 28-05-2024
to see stock.
Description
Description
  • Combi-Clip is included for fastening the evaporating flask
Specifications
Specifications
Rotavapor R-300
Vapor Duct
Glass Assembly V, C, S, E, CR, BY, HP
Borosilicate Glass 3.3
Combi-Clip
24/40
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers