Learn More
Merck Sampling Capsule for Metals Analysis, 1.0 μm
Disposable, EPA-accepted method for collecting groundwater prior to dissolved metals analysis.
€ 71.40 - € 620.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | Certified for 67 metals and 2 anions; meet U.S. EPA filtration requirements for 0.45 μm filtration. |
Pore Size | 1.0 μm |
Sterility | Non-sterile |
Filtration Area | 600 cm2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10275833
|
Merck
GWSC10001 |
1Pack |
€ 71.40
Pack of 50 |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285833
|
Merck
GWSC10010 |
10Pack |
€ 620.00
Pack of 10 |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Disposable, EPA-accepted method for collecting groundwater prior to dissolved metals analysis.
These groundwater sampling capsules are an EPA-approved method for collection of groundwater prior to dissolved metal analysis. The pleated membrane provides four times more filtration area than a 142 mm filter disc for faster flow rates and higher throughput.Features & Benefits:
- Each capsule includes a certificate indicating lowest detectable level (LDL) analysis for 67 metals and 2 anions
- Low-extractable filter and housing materials minimize background during sample analysis
- Available with three different pore sizes to suit a variety of water conditions: 5.0 μm for high particulate levels, 1.0 μm for normal particulate levels, 0.45 μm for relatively clean water
Applications: Environmental Monitoring, Dissolved Metals Analysis in Water
Specifications
Not Autoclavable | |
1.0 μm | |
600 cm2 | |
HDPE | |
7.6 cm |
Certified for 67 metals and 2 anions; meet U.S. EPA filtration requirements for 0.45 μm filtration. | |
Non-sterile | |
For metals analysis | |
Pharmaceutical Filters or Ultrafilters |