Learn More
Robu™ Series 16 2 Porosity Biplane Filter Disc
Brand:  Robu™ 62002
Additional Details : Weight : 0.10000kg
Product Code. 13477347
  • € 422.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Specifications
1Pack
Filter Disc
11.5 mm
200 mm