Learn More
Nabertherm™ Split Type Horizontal Tube Furnace RSH 1300 Series with B500 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
€ 5510.00 - € 13380.00
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17398149
|
Nabertherm™
RS152L3AN |
50 mm | 250 mm |
€ 5510.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17318159
|
Nabertherm™
RS252L3AN |
50 mm | 500 mm |
€ 6210.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17358159
|
Nabertherm™
RS322L3AN |
120 mm | 500 mm |
€ 8080.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17398159
|
Nabertherm™
RS382L3AN |
80 mm | 500 mm |
€ 7810.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17338169
|
Nabertherm™
RS422L3AN |
120 mm | 750 mm |
€ 10640.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17378169
|
Nabertherm™
RS472L3AN |
170 mm | 750 mm |
€ 11830.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17318179
|
Nabertherm™
RS482L3AN |
80 mm | 750 mm |
€ 10210.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17358179
|
Nabertherm™
RS522L3AN |
120 mm | 1000 mm |
€ 11720.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
17398179
|
Nabertherm™
RS572L3AN |
170 mm | 1000 mm |
€ 13380.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1300°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type S thermocouple
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 including fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- Switchgear and control unit integrated in furnace housing
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
B500 | |
Up to 1300°C |