Learn More
B Braun™ CUTFIX Stainless Steel Disposable Scalpels
Protects blades and prevents injuries with plastic cover while not in use. B Braun™ Aesculap™ Stainless Steel Disposable Scalpels include a plastic handle and blade protector. Sterile.
€ 8.15 - € 10.25
Specifications
Blade Material | Stainless Steel |
---|---|
Disposable | Yes |
Sterility | Sterile |
Handle Material | Plastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10354311
|
B Braun™
5518067 |
# 20 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10242611
|
B Braun™
5518016 |
# 23 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Sterile scalpels with plastic handle and blade protector.- Blades constructed of Stainless Steel.
- Disposable
Specifications
Stainless Steel | |
Yes | |
Sterile | |
Plastic |