missing translation for 'onlineSavingsMsg'
Learn More

Wuertt Celluloid™ Stainless Steel Test Tube Rack

€ 17.26 - € 113.42

Specifications

Product Type Test Tube Rack
Temperature (Metric) Range 80°C
Height (Metric) 70 mm
View More Specs

Products 3
Product Code Brand Array Price Quantity & Availability  
Product Code Brand Array Price Quantity & Availability  
10529651
View Documents Promotion Details
Wuertt Celluloid™
527101015
10 x 10
€ 17.26
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10692771
View Documents Promotion Details
Wuertt Celluloid™
52721213
2 x 12
€ 113.42
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
 
10485231
View Documents Promotion Details
Wuertt Celluloid™
52731213
3 x 12 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Test Tube Rack
80°C
70 mm
Videos
SDS
Documents

Documents

Product Certifications