Learn More
Medline™ Technico™ DIGIT-090 Pocket Refractometer
Brand:  Medline™ 8100.0900M
Additional Details : Weight : 1.00000kg
Product Code. 10313295
  • € 865.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Specifications
Pocket Refractometer
DIGIT-090
1 Each
0 to 42 Brix, 42 to 71 Brix, 71 to 90 Brix
Zero calibration with distilled water at 20°