Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint Without Graduation
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05700242
Additional Details : Weight : 0.50000kg
Description
- With round bottom
- With NS socket
- Available without stopper
- With PE stopper or with hexagonal hollow glass stopper
- Made of DURAN™ tubing
Specifications
Hexagonal glass stopper | |
Borosilicate Glass (3.3) | |
200 mm | |
28 mm |