Learn More
Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Foam brick ice packs never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control. Refrigerant bricks are vacuum-sealed for consistent weight and shape.
Brand: Sonoco™ FPP18
Additional Details : Weight : 13.50000kg
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
Specifications
28 Cs. | |
8.25 in. | |
20.9 cm | |
Refrigerant Ice Pack | |
18 oz. | |
8.25 x 10 x 0.75 in. (20.9 x 20.3 x 1.9 cm) |
14 cm | |
5.5 in. | |
0.75 in. | |
1.9 cm | |
0.72 kg | |
0°C (32°F) |