missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCA Hygiene Products™ Tork™ Xpress™ Multifold Hand Towel
Brand: SCA Hygiene Products™ 100297
Additional Details : Weight : 6.66000kg
Specifications
Multi-fold Hand Towel | |
21.2 cm | |
White |
34 cm | |
FSC | |
2-ply |