Learn More
IKA™ TR 4 Tube Insert
Brand:  IKA™ 0004525400
Additional Details : Weight : 0.10000kg
Product Code. 14871461
  • € 54.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Specifications
TR 4 Tube Insert
IKA Trayster Basic and Trayster Digital, Suitable 6 x 50mL Tubes with 28mm dia.
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers