Learn More
Idex Low Pressure Y Connectors
Combine, split, or intersect fluid streams throughout your system
Brand: Idex 02013-91
Additional Details : Weight : 0.25000kg
Description
Combine, split, or intersect fluid streams throughout your system
- Manufactured from inert polymers
Use Y connectors in a variety of low-pressure situations throughout your fluidic system. Y connectors create less turbulence and solvent outgassing than Tee connectors, improving analytical results. Handy mounting holes make it a snap to mount connectors. Y connector assemblies come complete with nuts and ferrules.
Specifications
Low Pressure Y Connector | |
1000 psi (69 bar) | |
P-235/P-200 fittings | |
Split a Stream or Join Two Streams Together |
0.02 in. | |
17 μL | |
PEEK |