Learn More
Kinesis™ Precolumn Inline Filters
Protect larger columns and injections from harmful particulates
Brand:  Idex 02011-00
Additional Details : Weight : 1.81440kg
 View more versions of this product
Product Code. 12936259
  • € 165.00 / Each
Estimated Shipment: 21-10-2024
to see stock.
Add to basket
Description
Description
  • Ideal for higher-flow systems
  • Interchangeable frits make protection more economical
  • Compatible with SFC and HPLC
  • Available to fit a variety of tubing sizes
Specifications
Specifications
2 μm
Precolumn Inline Filter
0.94 in.
Stainless Steel
0.66 uL