Learn More
Edwards Vacuum Rotary Vane Vacuum Pump Exhaust Oil Mist Filters
Captures oil mist from pump outlet when used with RV Rotary Vane Pumps
€ 526.79 - € 633.03
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
10034970
|
Edwards
A46229000 |
Model RV12, E2M18 pump |
€ 526.79
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12705058
|
Edwards
A46226000.. |
Model RV3, RV5, RV8 pump and EMF10 mist filter |
€ 633.03
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
Captures oil mist from pump outlet when used with RV Rotary Vane Pumps
Ejection of oil into the atmosphere during usage of gas ballast or pump high gas throughputs can be prevented.
- Semi-transparent white bottom half for oil level monitoring
- Integral pressure relief valve opens if oil element is blocked
- Neutralize oil mist smell
- Integral pressure relief valve opens if oil element blocks
- Efficient at 99.999% DOP test
- Azide proof
Use optional oil return kits for reduce oil level maintenance and savings in pump oil
Certifications
Meets COSHH and health and safety regulations
Specifications
Oil Mist Filter |