missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ UniPrep™ Syringeless Filters - Glass Microfiber

Process samples in one-third the time of conventional syringe filters

Brand:  Cytiva UN113UGMF

Additional Details : Weight : 0.40800kg

Product Code. 11338724

  • € 287.00 / Pack of 50
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

UniPrep syringeless filters are preassembled filtration devices for the filtration and storage of laboratory samples.

  • Layered glass microfiber depth filter for use with samples containing aqueous or organic solvents (indicated pore size is the particle retention rating)
  • Consists of external sample test tube, plunger (which incorporates membrane filter and prefilter), removable cap
  • Filtration area: 0.3cm2
  • Safe, efficient—prefilter and membrane can't pop off
  • Air purge unnecessary—inserting plunger into tube provides a positive pressure purge of sample through filter
  • Sealed glass microfiber prefilter traps larger particles before they can clog the membrane

  • Increase lab productivity and efficiency
  • Generate less waste
  • Realize greater overall cost savings per test

Specifications

Specifications

Glass Microfiber
0.28 cm2
Syringeless Filter
0.45 μm
50 μL
17 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers