Contec™ PROSAT™ PS-911 Presaturated Meltblown Polypropylene Wipes
PROSAT™ wipes are an effective, efficient and convenient method of cleaning process tools and other equipment. PROSAT wipes are also ideal for wiping articles prior to pass through.
Description
497.49 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 11.59200kg Transport: UN number : 3175 Chem class : 4.1 Pack group : II
Product Code. 17264513
Quantity Price
1 €  782.0 / Pack of 1500
In stock, ready to ship:
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
Description and Specification
Specification
Product Type Pre-saturated Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 11 in.
Width (Metric) 28 cm
Material Polypropylene
Certifications/Compliance ISO Class 5, SEMI for metals/contaminants
Disposable Single-use
Dimensions (L x W) 23 x 28 cm (9 x 11 in.)
For Use With (Application) Ideal for Routine Cleaning and Wiping Instruments and Equipment
For Use With (Equipment) Process Tools and Other Equipment
Packaging Quantity 30 Pk., 50 Pk./Cs.
Saturation Solution 70% IPA/ 30% DI Water
Sterility Non-sterile
  • Presaturated with IPA and DI water blends in flat, resealable pouches individually lot-numbered for traceability
  • These specially designed pouches preserve cleanliness and solvent saturation levels
  • Solvent blend meets SEMI specifications for metals and contaminants
  • ISO Class 5 compatible
  • 30 wipes/pouch, 50 pouches/case
Alternative Products
Safety and Handling
[CONTEC PROSAT MBPP WITH 70% IPA]
[Danger]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H335, sequence=3, description=May cause respiratory irritation.}, {code=H336, sequence=4, description=May cause drowsiness or dizziness.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P243, sequence=52, description=Take precautionary measures against static discharge.}, {code=P261, sequence=53, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P270, sequence=54, description=Do not eat, drink or smoke when using this product.}, {code=P280, sequence=55, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=56, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=57, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=58, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=59, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P337+P313, sequence=60, description=If eye irritation persists: Get medical advice/attention.}, {code=P370+P378, sequence=61, description=In case of fire: Use to extinguish.}, {code=P403+P233, sequence=62, description=Store in a well-ventilated place. Keep container tightly closed.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: propan-2-ol}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
  • CONTEC PROSAT MBPP WITH 70% IPA
Signal Word
  • Danger
Hazard Statement
  • H225- Highly flammable liquid and vapour.
  • H319- Causes serious eye irritation.
  • H335- May cause respiratory irritation.
  • H336- May cause drowsiness or dizziness.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243- Take precautionary measures against static discharge.
  • P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270- Do not eat, drink or smoke when using this product.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312- Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313- If eye irritation persists: Get medical advice/attention.
  • P370+P378- In case of fire: Use to extinguish.
  • P403+P233- Store in a well-ventilated place. Keep container tightly closed.
Supplemental Information
  • MIXTURE LIST- Contain: propan-2-ol