151.32 EUR valid until 2025-12-31
Use promo code "24367" to get your promotional price.
Use promo code "24367" to get your promotional price.
| Certifications/Compliance | ISO 9001:2015, FDA 21 CFR, GMP |
|---|---|
| Color | Clear |
| Volume (Metric) | 0.2 mL |
| Quantity | 120 Strips/Bag |
| For Use With (Equipment) | PCR Strip Tubes with 8 Wells for Most Thermocyclers |
| Product Line | PurePlus |
| Sterility | Non-sterile |
| Material | Polypropylene USP Class VI Resin |
| Closure Type | Domed Cap (Attached) |
| No. of Wells | 8 |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Product Type | PCR Tube |