accessibility menu, dialog, popup

UserName

CAS RN 1011-16-1

IUPAC Name: 1-(2-fluorophenyl)piperazine hydrochloride
Synonyms: 1-(2-fluorophenyl)piperazine hydrochloride
Molecular Weight (g/mol): 216.68
Molecular Formula: C10H14ClFN2
InChi Key: JVQOFQHGEFLRRQ-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=CC=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results