accessibility menu, dialog, popup

UserName

CAS RN 101477-54-7

HHClClH3CONNFFOH3COCH3

CAS RN 101477-54-7

IUPAC Name: dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine dichloride
Synonyms: dihydrogen lomerizine dichloride
Molecular Weight (g/mol): 541.46
Molecular Formula: C27H32Cl2F2N2O3
InChi Key: LOGVKVSFYBBUAJ-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC=C(CN2CCN(CC2)C(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)C(OC)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results