CAS RN 10151-46-9
CAS RN 10151-46-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Naphthalene-2-sulfonohydrazide, TRC
CAS: 10151-46-9 Molecular Formula: C10 H10 N2 O2 S Molecular Weight (g/mol): 222.264 Synonym: (Naphthalen-2-yl)sulfonyl Hydrazine,(beta-Naphthylsulfonyl)hydrazine,2-Naphthalenesulfonyl Hydrazide,2-Naphthylsulfonyl Hydrazide2-Naphthylsulfonyl Hydrazine IUPAC Name: naphthalene-2-sulfonohydrazide SMILES: NNS(=O)(=O)c1ccc2ccccc2c1