CAS RN 101565-95-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
meta-Methoxytopolin Riboside, TRC
CAS: 101565-95-1 Molecular Formula: C18H21N5O5 Molecular Weight (g/mol): 387.39 Synonym: N-[(3-Methoxyphenyl)methyl]adenosine,MemTR,6-(3-Methoxybenzylamino)-9-ß-dribofuranosylpurine,N-[(3-Methoxyphenyl)methyl]-adenosine IUPAC Name: (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(3-methoxyphenyl)methylamino]purin-9-yl]oxolane-3,4-diol SMILES: COc1cccc(CNc2ncnc3c2ncn3[C@@H]4O[C@H](CO)[C@H](O)[C@@H]4O)c1