accessibility menu, dialog, popup

UserName

CAS RN 10166-09-3

CH3OCl

CAS RN 10166-09-3

IUPAC Name: 2-(2-methylphenyl)acetyl chloride
Synonyms: 2-(2-methylphenyl)acetyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: FKMIBYMTHOJWCY-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CC(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results