CAS RN 1016636-76-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido-[2,3-d]pyrimidin-7(8H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido-[2,3-d]pyrimidin-7(8H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido-[2,3-d]pyrimidin-7(8H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.