accessibility menu, dialog, popup

UserName

CAS RN 10192-30-0

NH3OHSOHO

CAS RN 10192-30-0

IUPAC Name: sulfurous acid amine
Synonyms: sulfurous acid amine
Molecular Weight (g/mol): 99.1
Molecular Formula: H5NO3S
InChi Key: ZETCGWYACBNPIH-UHFFFAOYSA-N
SMILES: N.OS(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results