CAS RN 101931-00-4
CAS RN 101931-00-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Pioglitazone (M-II) (Mixture of Diastereomers), TRC
CAS: 101931-00-4 Molecular Formula: C19 H20 N2 O4 S Molecular Weight (g/mol): 372.44 Synonym: 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)-2-hydroxyethoxy]phenyl]methyl]-,5-[[4-[2-(5-Ethyl-2-pyridyl)-2-hydroxy-ethoxy]phenyl]methyl]thiazolidine-2,4-dione IUPAC Name: 5-[[4-[2-(5-ethylpyridin-2-yl)-2-hydroxyethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione SMILES: CCc1ccc(nc1)C(O)COc2ccc(CC3SC(=O)NC3=O)cc2