accessibility menu, dialog, popup

UserName

CAS RN 102-77-2

IUPAC Name: 2-(morpholin-4-ylsulfanyl)-1,3-benzothiazole
Synonyms: 2-(morpholin-4-ylsulfanyl)-1,3-benzothiazole
Molecular Weight (g/mol): 252.35
Molecular Formula: C11H12N2OS2
InChi Key: MHKLKWCYGIBEQF-UHFFFAOYSA-N
SMILES: C1CN(CCO1)SC1=NC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)