CAS RN 1020719-27-0
CAS RN 1020719-27-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-hydroxyphenyl]-2-phenyl-1-butene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.