CAS RN 102212-26-0
CAS RN 102212-26-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ARN2966, MedChemExpress
MedChemExpress ARN2966 is a potent post-transcriptional modulator of APP expression; reduces expression of APP with resultant lower production of A(beta).

