CAS RN 1028327-66-3
IUPAC Name:
4-(4-methylpiperazin-1-yl)-6-phenylpyrimidin-2-amine
Synonyms:
4-(4-methylpiperazin-1-yl)-6-phenylpyrimidin-2-amine
Molecular Weight (g/mol):
269.35
Molecular Formula:
C15H19N5
InChi Key:
NIJGWJIOMPHDBP-UHFFFAOYSA-N
SMILES:
CN1CCN(CC1)C1=NC(N)=NC(=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
VUF10460, MedChemExpress
MedChemExpress VUF10460 is a non-imidazole histamine H4 receptor agonist; binds to rat H4 receptor with a pKi of 7.46.