CAS RN 102839-00-9
CAS RN 102839-00-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2,2'-(1,4-Piperazinediyl)bis[6,7-dimethoxy-4-quinazolinamine] (Terazosin Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2'-(1,4-Piperazinediyl)bis[6,7-dimethoxy-4-quinazolinamine] (Terazosin Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2'-(Piperazin-1,4-diyl)bis(6,7-dimethoxyquinazolin-4-amine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.