CAS RN 1029890-89-8
CAS RN 1029890-89-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-[(4R,5R)-4,5-Dihydroxy-L-ornithine]-Echinocandin B Hydrochloride, TRC
CAS: 1029890-89-8 Molecular Formula: C34H51N7O15 . HCl Molecular Weight (g/mol): 834.3 Synonym: Anidulafungin Nucleus SMILES: Cl.C[C@@H](O)[C@@H]1NC(=O)C(NC(=O)[C@@H]2C[C@@H](O)CN2C(=O)C(NC(=O)[C@@H](N)C[C@@H](O)[C@@H](O)NC(=O)[C@@H]3[C@@H](O)[C@@H](C)CN3C1=O)[C@@H](C)O)[C@H](O)[C@@H](O)c4ccc(O)cc4