accessibility menu, dialog, popup

Test

CAS RN 103300-74-9

CAS RN 103300-74-9

IUPAC Name: (4S)-N-[(2S)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-(1H-imidazol-5-yl)-1-oxopropan-2-yl]-1-methyl-2,6-dioxo-1,3-diazinane-4-carboxamide
Synonyms: taltirelin
Molecular Weight (g/mol): 405.42
Molecular Formula: C17H23N7O5
InChi Key: LQZAIAZUDWIVPM-SRVKXCTJSA-N
SMILES: CN1C(=O)C[C@H](NC1=O)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N1CCC[C@H]1C(N)=O
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)