CAS RN 1033805-22-9
CAS RN 1033805-22-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Telotristat Ethyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Telotristat Ethyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Telotristat Ethyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Telotristat ethyl, MedChemExpress
MedChemExpress Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.