CAS RN 1035555-63-5
CAS RN 1035555-63-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TAK-733, MedChemExpress
MedChemExpress TAK-733 is a potent and selective MEK allosteric site inhibitor with an IC50 of 3.2 nM.

