CAS RN 1037624-75-1
CAS RN 1037624-75-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bemcentinib, MedChemExpress
MedChemExpress Bemcentinib (R428) is a potent and selective inhibitor of Axl with an IC50 of 14 nM.

